Assay Details
Gene Symbol: Ak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033967.2 | NP_001029139.1 | 2 - 3 | 283 | 66 | |
NM_030986.2 | NP_112248.2 | 2 - 3 | 283 | 66 | ||
GenBank mRNA | BC061727.1 | - | 2 - 3 | 237 | 66 | |
CO570724.1 | - | 2 - 3 | 283 | 66 | ||
D13061.1 | - | 2 - 3 | 250 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
