Assay Details
Gene Symbol: Ap2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080583.1 | NP_542150.1 | 13 - 14 | 1937 | 70 | |
XM_008767935.1 | XP_008766157.1 | 14 - 15 | 2694 | 70 | ||
XM_008767936.1 | XP_008766158.1 | 13 - 14 | 1908 | 70 | ||
XM_008767937.1 | XP_008766159.1 | 12 - 13 | 1823 | 70 | ||
XM_008767938.1 | XP_008766160.1 | 12 - 13 | 1840 | 70 | ||
XM_008767939.1 | XP_008766161.1 | 13 - 14 | 2676 | 70 | ||
GenBank mRNA | BC103481.1 | - | 14 - 15 | 1906 | 70 | |
M34176.1 | - | 13 - 14 | 1869 | 70 | ||
M77246.1 | - | 13 - 14 | 1937 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
