Assay Details
Gene Symbol: Mat2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134351.1 | NP_599178.1 | 2 - 3 | 243 | 86 | |
GenBank mRNA | BC062394.1 | - | 276 | 5622019 | 86 | |
J05571.1 | - | 2 - 3 | 243 | 86 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
