Assay Details
Gene Symbol: Aurka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153296.2 | NP_695208.2 | 4 - 5 | 558 | 62 | |
XM_006235675.2 | XP_006235737.1 | 5 - 6 | 637 | 62 | ||
XM_006235676.2 | XP_006235738.1 | 5 - 6 | 654 | 62 | ||
GenBank mRNA | AF537333.1 | - | 4 - 5 | 546 | 62 | |
BC104676.1 | - | 4 - 5 | 558 | 62 | ||
FQ227101.1 | - | 5 - 6 | 644 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
