Assay Details
Gene Symbol: Cox7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182819.2 | NP_877971.2 | 2 - 3 | 228 | 84 | |
GenBank mRNA | AY339885.1 | - | 2 - 3 | 188 | 84 | |
BC079123.1 | - | 2 - 3 | 228 | 84 | ||
FQ210062.1 | - | 2 - 3 | 223 | 84 | ||
FQ213156.1 | - | 2 - 3 | 259 | 84 | ||
FQ217174.1 | - | 2 - 3 | 253 | 84 | ||
FQ217253.1 | - | 2 - 3 | 246 | 84 | ||
FQ217570.1 | - | 2 - 3 | 259 | 84 | ||
FQ217933.1 | - | 2 - 3 | 236 | 84 | ||
FQ217995.1 | - | 2 - 3 | 258 | 84 | ||
FQ218015.1 | - | 2 - 3 | 258 | 84 | ||
FQ218434.1 | - | 2 - 3 | 258 | 84 | ||
FQ221598.1 | - | 2 - 3 | 258 | 84 | ||
FQ221667.1 | - | 2 - 3 | 257 | 84 | ||
FQ222019.1 | - | 2 - 3 | 257 | 84 | ||
FQ222086.1 | - | 2 - 3 | 246 | 84 | ||
FQ222576.1 | - | 2 - 3 | 250 | 84 | ||
FQ223858.1 | - | 2 - 3 | 246 | 84 | ||
FQ224255.1 | - | 2 - 3 | 246 | 84 | ||
FQ224424.1 | - | 2 - 3 | 258 | 84 | ||
FQ224448.1 | - | 2 - 3 | 258 | 84 | ||
FQ225820.1 | - | 2 - 3 | 258 | 84 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
