Assay Details
Gene Symbol: Mobp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012720.1 | NP_036852.1 | 3 - 4 | 319 | 86 | |
GenBank mRNA | BC087694.1 | - | 2 - 3 | 273 | 86 | |
X87900.1 | - | 3 - 4 | 312 | 86 | ||
X89637.1 | - | 3 - 4 | 319 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P074471 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P167930 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377665 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654610 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266366 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625620 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948403 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950028 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450120 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359771 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908857 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908930 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564006 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193301 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64320 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945256 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157018 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75313 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P167930 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P134953 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P074471 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P134953 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P167930 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P074471 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P134953 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P074471 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P167930 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P167930 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P245674 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P353699 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701749945 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700767040 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 1741 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 89511 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 83182 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 701749945 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700767040 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10914322 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10914319 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10914324 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10914328 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10914330 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10914320 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10914323 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10914325 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10914327 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10914326 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10914329 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10914318 | - | - |
Rat Genome 230 2.0 Array | 1368263_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1370434_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1370500_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1370500_a_at | - | - |
Rat Genome 230 2.0 Array | 1370434_a_at | - | - |
Rat Genome 230 2.0 Array | 1368263_a_at | - | - |
Affymetrix HT Rat Focus Array | 1370434_a_at | - | - |
Affymetrix HT Rat Focus Array | 1368263_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1370500_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1370500_a_at | - | - |
Affymetrix HT Rat Focus Array | 1368263_a_at | - | - |
Affymetrix HT Rat Focus Array | 1370434_PM_a_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0800013994.rn.1 | TC0800001757.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0800013995.rn.1 | TC0800001757.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0800013998.rn.1 | TC0800001757.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0800013990.rn.1 | TC0800001757.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0800013997.rn.1 | TC0800001757.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0800013996.rn.1 | TC0800001757.rn.1 | RTA TC |
Rat Expression Set 230 | 1370434_a_at | - | - |
Rat Expression Set 230 | 1370500_a_at | - | - |
Rat Expression Set 230 | 1370434_PM_a_at | - | - |
Rat Expression Set 230 | 1368263_PM_a_at | - | - |
Rat Expression Set 230 | 1370500_PM_a_at | - | - |
Rat Expression Set 230 | 1368263_a_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10914318 | - | - |
Rat Genome U34 Set | D28110_at | - | - |
Rat Genome U34 Set | D28111_at | - | - |
Rat Genome U34 Set | D28111_g_at | - | - |
Rat Genome U34 Set | D28110_g_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20792962 | - | - |
