Assay Details
Gene Symbol: Zfand5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106356.1 | NP_001099826.1 | 1 - 2 | 656 | 129 | |
XM_008760297.1 | XP_008758519.1 | 3 - 4 | 855 | 129 | ||
GenBank mRNA | BC168882.1 | - | 3 - 4 | 323 | 129 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001106356.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
