Assay Details
Gene Symbol: Samd4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107498.1 | NP_001100968.1 | 11 - 12 | 2238 | 70 | |
XM_008759143.1 | XP_008757365.1 | 12 - 13 | 2361 | 70 | ||
GenBank mRNA | BC090319.1 | - | 12 - 13 | 2176 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P012647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683332 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849421 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538082 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211652 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707378 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427457 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628599 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397365 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810234 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14508 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87338 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164565 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P012647 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P012647 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P012647 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P515577 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 90750 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 65280 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10720232 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10720227 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10720226 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10720228 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10720230 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10720225 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10720235 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10720236 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10720233 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10720234 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10720229 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10720231 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10720224 | - | - |
Rat Genome 230 2.0 Array | 1386856_a_at | - | - |
Rat Genome 230 2.0 Array | 1386854_PM_at | - | - |
Rat Genome 230 2.0 Array | 1386854_at | - | - |
Rat Genome 230 2.0 Array | 1386856_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1394105_PM_at | - | - |
Rat Genome 230 2.0 Array | 1394105_at | - | - |
Affymetrix HT Rat Focus Array | 1386856_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1386854_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1394105_at | - | - |
Affymetrix HT Rat Focus Array | 1386856_a_at | - | - |
Affymetrix HT Rat Focus Array | 1394105_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1386854_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100008606.rn.1 | TC0100001159.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100008607.rn.1 | TC0100001159.rn.1 | RTA TC |
Rat Expression Set 230 | 1394105_PM_at | - | - |
Rat Expression Set 230 | 1386854_at | - | - |
Rat Expression Set 230 | 1386854_PM_at | - | - |
Rat Expression Set 230 | 1386856_a_at | - | - |
Rat Expression Set 230 | 1394105_at | - | - |
Rat Expression Set 230 | 1386856_PM_a_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10720224 | - | - |
Rat Genome U34 Set | rc_AA859633_at | - | - |
Rat Genome U34 Set | rc_AA859633_g_at | - | - |
Rat Genome U34 Set | rc_AA850505_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21908234 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365617 | - | - |
