Assay Details
Gene Symbol: Eif4e2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC158817.1 | - | 689 | 102 | ||
FQ220431.1 | - | 805 | 102 | |||
FQ226325.1 | - | 774 | 102 | |||
FQ231706.1 | - | 774 | 102 | |||
FQ232529.1 | - | 774 | 102 | |||
FQ233479.1 | - | 710 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
