Assay Details
Gene Symbol: Rspo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130575.1 | NP_001124047.1 | 1 - 2 | 1088 | 71 | |
XM_008765455.1 | XP_008763677.1 | 1 - 2 | 537 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
osteoblast differentiation
biological_process
bone mineralization
embryonic forelimb morphogenesis
embryonic hindlimb morphogenesis
negative regulation of odontogenesis of dentin-containing tooth
lung growth
epithelial tube branching involved in lung morphogenesis
trachea cartilage morphogenesis
dopaminergic neuron differentiation
positive regulation of canonical Wnt signaling pathway
G-protein coupled receptor binding
molecular_function
receptor binding
heparin binding
osteoblast differentiation
biological_process
bone mineralization
embryonic forelimb morphogenesis
embryonic hindlimb morphogenesis
negative regulation of odontogenesis of dentin-containing tooth
lung growth
epithelial tube branching involved in lung morphogenesis
trachea cartilage morphogenesis
dopaminergic neuron differentiation
positive regulation of canonical Wnt signaling pathway
G-protein coupled receptor binding
molecular_function
receptor binding
heparin binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P082919 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P082924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641665 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538730 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356532 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468850 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251658 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855717 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228339 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412702 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810492 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682124 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758903 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135579 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52827 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286244 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94837 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520584 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947465 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910290 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263156 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454538 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120293 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584795 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147454 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504021 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918159 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357222 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362247 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769274 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197747 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929559 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89307 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287174 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423290 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701545 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645357 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964580 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715494 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730320 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378425 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686992 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555589 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573789 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603390 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226197 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214132 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179730 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30285 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957677 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289082 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32630 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123992 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54067 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15458 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97366 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140338 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23598 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P082924 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P082919 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P082919 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P082924 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P082919 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P082924 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P610634 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10903540 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903542 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903539 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903543 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903541 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903544 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10903538 | - | - |
Affymetrix Rat Clariom S Assay | PSR0700023676.rn.1 | TC0700003070.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700023674.rn.1 | TC0700003070.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10903538 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21987901 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1368734 | - | - |
