Assay Details
Gene Symbol: Dapk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013109.1 | NP_001013127.1 | 1 - 2 | 216 | 92 | |
XM_006243306.2 | XP_006243368.1 | 5 - 6 | 733 | 92 | ||
XM_006243307.1 | XP_006243369.1 | 2 - 3 | 210 | 92 | ||
XM_006243308.2 | XP_006243370.1 | 2 - 3 | 1412 | 92 | ||
XM_006243309.2 | XP_006243371.1 | 2 - 3 | 1107 | 92 | ||
XM_008766241.1 | XP_008764463.1 | 5 - 6 | 743 | 92 | ||
GenBank mRNA | BC091410.1 | - | 1 - 2 | 216 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
