Assay Details
Gene Symbol: Casp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022522.2 | NP_071967.2 | 1 - 2 | 81 | 59 | |
GenBank mRNA | AF136231.1 | - | 1 - 2 | 79 | 59 | |
AF136232.1 | - | 1 - 2 | 79 | 59 | ||
U77933.1 | - | 1 - 2 | 81 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
