Assay Details
Gene Symbol: Psmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033236.1 | NP_150239.1 | 9 - 10 | 860 | 94 | |
GenBank mRNA | BC061542.1 | - | 9 - 10 | 902 | 94 | |
D50694.1 | - | 9 - 10 | 860 | 94 | ||
FQ213265.1 | - | 6 - 7 | 663 | 94 | ||
FQ215767.1 | - | 6 - 7 | 663 | 94 | ||
FQ225383.1 | - | 6 - 7 | 663 | 94 | ||
FQ230973.1 | - | 9 - 10 | 918 | 94 | ||
FQ234824.1 | - | 6 - 7 | 663 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
