Assay Details
Gene Symbol: Fgf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130814.1 | NP_570827.1 | 3 - 4 | 418 | 114 | |
GenBank mRNA | AB079374.1 | - | 3 - 4 | 418 | 114 | |
AF348446.1 | - | 2 - 3 | 232 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
