Assay Details
Gene Symbol: Rad18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077673.1 | NP_001071141.1 | 2 - 3 | 176 | 121 | |
XM_006237036.1 | XP_006237098.1 | 2 - 3 | 205 | 121 | ||
XM_006237037.1 | XP_006237099.1 | 2 - 3 | 205 | 121 | ||
GenBank mRNA | BC127510.1 | - | 2 - 3 | 176 | 121 | |
EF395817.1 | - | 146 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
