Assay Details
Gene Symbol: Tia1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012096.1 | NP_001012096.1 | 1 - 2 | 203 | 77 | |
XM_006236828.2 | XP_006236890.1 | 1 - 2 | 277 | 77 | ||
XM_006236829.2 | XP_006236891.1 | 1 - 2 | 279 | 77 | ||
XM_006236830.2 | XP_006236892.1 | 1 - 2 | 277 | 77 | ||
GenBank mRNA | BC087064.1 | - | 1 - 2 | 203 | 77 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC087064.1, NM_001012096.1, XM_006236828.2, XM_006236829.2, XM_006236830.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
