Assay Details
Gene Symbol: Adipor2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006237183.1 | XP_006237245.1 | 7 - 8 | 1344 | 68 | |
XM_006237184.2 | XP_006237246.1 | 7 - 8 | 1215 | 68 | ||
XM_006237185.2 | XP_006237247.1 | 7 - 8 | 1230 | 68 | ||
XM_006237186.2 | XP_006237248.1 | 7 - 8 | 1359 | 68 | ||
GenBank mRNA | AY724514.1 | - | 7 - 8 | 1298 | 68 | |
FQ232508.1 | - | 7 - 8 | 1317 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
