Assay Details
Gene Symbol: Sec13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006978.1 | NP_001006979.1 | 5 - 6 | 477 | 66 | |
GenBank mRNA | BC084705.1 | - | 5 - 6 | 477 | 66 | |
FQ212944.1 | - | 5 - 6 | 497 | 66 | ||
FQ216715.1 | - | 5 - 6 | 495 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
