Assay Details
Gene Symbol: Rnf185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024271.1 | NP_001019442.1 | 5 - 6 | 509 | 80 | |
XM_006251333.1 | XP_006251395.1 | 5 - 6 | 477 | 80 | ||
XM_008770331.1 | XP_008768553.1 | 3 - 4 | 321 | 80 | ||
GenBank mRNA | BC092655.1 | - | 5 - 6 | 509 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
