Assay Details
Gene Symbol: Ccdc24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134626.1 | NP_001128098.1 | 2 - 3 | 376 | 91 | |
XM_006238709.2 | XP_006238771.1 | 2 - 3 | 587 | 91 | ||
XM_006238710.2 | XP_006238772.1 | 2 - 3 | 575 | 91 | ||
XM_008764052.1 | XP_008762274.1 | 2 - 3 | 587 | 91 | ||
XM_008764053.1 | XP_008762275.1 | 2 - 3 | 575 | 91 | ||
XM_008764054.1 | XP_008762276.1 | 2 - 3 | 587 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
