Assay Details
Gene Symbol: Fam184a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256500.2 | XP_006256562.1 | 13 - 14 | 2610 | 72 | |
XM_008773011.1 | XP_008771233.1 | 15 - 16 | 2799 | 72 | ||
XM_008773013.1 | XP_008771235.1 | 14 - 15 | 2652 | 72 | ||
XM_008773015.1 | XP_008771237.1 | 16 - 17 | 2942 | 72 | ||
XM_008773016.1 | XP_008771238.1 | 15 - 16 | 2900 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
