Assay Details
Gene Symbol: Tab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012062.1 | NP_001012062.1 | 4 - 5 | 1773 | 62 | |
XM_006227620.2 | XP_006227682.1 | 3 - 4 | 1803 | 62 | ||
GenBank mRNA | BC085788.1 | - | 4 - 5 | 1773 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
