Assay Details
Gene Symbol: Srsf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239222.2 | XP_006239284.1 | 5 - 6 | 628 | 66 | |
XM_006239223.2 | XP_006239285.1 | 5 - 6 | 626 | 66 | ||
GenBank mRNA | FQ227288.1 | - | 5 - 6 | 605 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
