Assay Details
Gene Symbol: Fbxw11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106993.1 | NP_001100463.1 | 6 - 7 | 730 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
protein dephosphorylation
protein ubiquitination
SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
protein destabilization
positive regulation of circadian rhythm
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of proteolysis
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
negative regulation of NIK/NF-kappaB signaling
ubiquitin-protein transferase activity
ubiquitin protein ligase activity
protein polyubiquitination
protein dephosphorylation
protein ubiquitination
SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
protein destabilization
positive regulation of circadian rhythm
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of proteolysis
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
negative regulation of NIK/NF-kappaB signaling
ubiquitin-protein transferase activity
ubiquitin protein ligase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149163 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285107 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756328 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149257 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676567 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703919 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880367 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764842 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212553 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468156 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296940 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551972 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971425 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15601 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444643 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641993 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79272 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115690 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932633 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691196 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311968 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277690 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193987 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273106 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272920 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899736 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125339 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506095 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10579 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781514 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799444 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596936 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819792 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77644 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256255 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234802 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502948 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372338 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375410 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727225 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306066 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943744 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306163 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714030 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569728 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202562 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256658 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422162 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353661 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374677 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65377 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67731 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33839 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112516 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87607 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57138 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P626104 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P626104 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P626104 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P626104 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P206973 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 38415 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 16080 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 6943 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10732709 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10732703 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10732700 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10732705 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10732707 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10732706 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10732708 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10732711 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10732701 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10732702 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10732699 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10732697 | - | - |
Rat Genome 230 2.0 Array | 1374412_PM_at | - | - |
Rat Genome 230 2.0 Array | 1374412_at | - | - |
Affymetrix HT Rat Focus Array | 1374412_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1374412_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1000003154.rn.1 | TC1000000318.rn.1 | RTA TC |
Rat Expression Set 230 | 1374412_at | - | - |
Rat Expression Set 230 | 1374412_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10732697 | - | - |
Rat Genome U34 Set | rc_AI009208_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21215962 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355074 | - | - |
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358124 | - | - |
