Assay Details
Gene Symbol: Ndrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011991.1 | NP_001011991.1 | 5 - 6 | 463 | 86 | |
XM_006241708.2 | XP_006241770.1 | 5 - 6 | 735 | 86 | ||
GenBank mRNA | AY500369.1 | - | 5 - 6 | 425 | 86 | |
BC081898.1 | - | 5 - 6 | 463 | 7110455 | 86 | |
FQ212154.1 | - | 5 - 6 | 475 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
