Assay Details
Gene Symbol: Rabgef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108333.1 | NP_001101803.1 | 2 - 3 | 578 | 69 | |
XM_008769150.1 | XP_008767372.1 | 3 - 4 | 669 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
