Assay Details
Gene Symbol: Itgbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017505.1 | NP_001017505.1 | 5 - 6 | 818 | 85 | |
GenBank mRNA | BC087076.1 | - | 5 - 6 | 818 | 7134947 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
