Assay Details
Gene Symbol: Gnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031037.2 | NP_112299.1 | 2 - 3 | 283 | 79 | |
GenBank mRNA | AF277892.1 | - | 1 - 2 | 64 | 79 | |
BC065579.1 | - | 2 - 3 | 283 | 79 | ||
U34959.1 | - | 1 - 2 | 55 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
