Assay Details
Gene Symbol: Zfand5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106356.1 | NP_001099826.1 | 3 - 4 | 881 | 102 | |
XM_008760297.1 | XP_008758519.1 | 5 - 6 | 1080 | 102 | ||
GenBank mRNA | BC168882.1 | - | 5 - 6 | 548 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
