Assay Details
Gene Symbol: Ankrd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008771402.1 | XP_008769624.1 | 3 - 4 | 5001 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
