Assay Details
Gene Symbol: Galt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013089.2 | NP_001013107.1 | 4 - 5 | 450 | 93 | |
XM_006238033.2 | XP_006238095.1 | 4 - 5 | 374 | 93 | ||
GenBank mRNA | BC098756.1 | - | 4 - 5 | 450 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
