Assay Details
Gene Symbol: Ubac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034937.2 | NP_001030109.2 | 3 - 4 | 406 | 97 | |
XM_006252509.2 | XP_006252571.1 | 3 - 4 | 480 | 97 | ||
GenBank mRNA | FM053767.1 | - | 3 - 4 | 334 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
