Assay Details
Gene Symbol: Arhgef9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023957.1 | NP_076447.1 | 8 - 9 | 1705 | 94 | |
GenBank mRNA | AJ250425.1 | - | 8 - 9 | 1705 | 94 | |
AJ302676.1 | - | 7 - 8 | 1525 | 94 |
Gene Symbol: LOC100912165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773295.1 | XP_008771517.1 | 1 - 2 | 441 | 94 | |
XM_008773296.1 | XP_008771518.1 | 444 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
