Assay Details
Gene Symbol: Itgb1bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108245.2 | NP_001101715.1 | 10 - 11 | 869 | 98 | |
GenBank mRNA | FQ215476.1 | - | 10 - 11 | 864 | 98 | |
FQ216044.1 | - | 10 - 11 | 868 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
