Assay Details
Gene Symbol: Itgb1bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108245.2 | NP_001101715.1 | 6 - 7 | 527 | 64 | |
GenBank mRNA | FM116382.1 | - | 6 - 7 | 486 | 64 | |
FM118664.1 | - | 6 - 7 | 511 | 64 | ||
FQ215476.1 | - | 6 - 7 | 522 | 64 | ||
FQ216044.1 | - | 6 - 7 | 526 | 64 | ||
FQ216100.1 | - | 6 - 7 | 503 | 64 | ||
FQ216185.1 | - | 6 - 7 | 523 | 64 | ||
FQ224925.1 | - | 6 - 7 | 524 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
