Assay Details
Gene Symbol: Actr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009268.1 | NP_001009268.1 | 5 - 6 | 651 | 92 | |
GenBank mRNA | BC088442.1 | - | 5 - 6 | 651 | 92 | |
FQ212547.1 | - | 5 - 6 | 650 | 92 | ||
FQ212814.1 | - | 6 - 7 | 615 | 92 | ||
FQ219071.1 | - | 5 - 6 | 600 | 92 | ||
FQ225424.1 | - | 5 - 6 | 622 | 92 | ||
FQ226083.1 | - | 5 - 6 | 597 | 92 | ||
FQ226422.1 | - | 5 - 6 | 600 | 92 | ||
FQ227830.1 | - | 5 - 6 | 600 | 92 | ||
FQ230292.1 | - | 1 - 2 | 143 | 92 | ||
FQ231828.1 | - | 5 - 6 | 602 | 92 | ||
FQ231874.1 | - | 5 - 6 | 635 | 92 | ||
FQ234230.1 | - | 5 - 6 | 602 | 92 | ||
FQ235071.1 | - | 5 - 6 | 622 | 92 | ||
FQ235201.1 | - | 5 - 6 | 617 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
