Assay Details
Gene Symbol: Pitpna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017231.1 | NP_058927.1 | 2 - 3 | 245 | 89 | |
GenBank mRNA | BC070945.1 | - | 2 - 3 | 273 | 7103390 | 89 |
M25758.1 | - | 2 - 3 | 245 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
