Assay Details
Gene Symbol: Eif5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033681.1 | NP_001028853.1 | 4 - 5 | 514 | 77 | |
XM_006246635.2 | XP_006246697.1 | 5 - 6 | 744 | 77 | ||
XM_006246637.2 | XP_006246699.1 | 5 - 6 | 712 | 77 | ||
XM_006246638.1 | XP_006246700.1 | 4 - 5 | 733 | 77 | ||
GenBank mRNA | BC101891.1 | - | 4 - 5 | 514 | 7930704 | 77 |
FQ212627.1 | - | 3 - 4 | 329 | 77 | ||
FQ213502.1 | - | 4 - 5 | 551 | 77 | ||
FQ219881.1 | - | 4 - 5 | 598 | 77 | ||
FQ220490.1 | - | 3 - 4 | 269 | 77 | ||
FQ220539.1 | - | 4 - 5 | 528 | 77 | ||
FQ226884.1 | - | 4 - 5 | 526 | 77 | ||
FQ227550.1 | - | 2 - 3 | 150 | 77 | ||
FQ229255.1 | - | 4 - 5 | 537 | 77 | ||
FQ231501.1 | - | 4 - 5 | 496 | 77 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
