Assay Details
Gene Symbol: Spns3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109035.1 | NP_001102505.1 | 3 - 4 | 593 | 77 | |
XM_006246786.2 | XP_006246848.1 | 4 - 5 | 1182 | 77 | ||
XM_006246789.2 | XP_006246851.1 | 4 - 5 | 1183 | 77 | ||
XM_008767857.1 | XP_008766079.1 | 4 - 5 | 1183 | 77 | ||
XM_008767858.1 | XP_008766080.1 | 4 - 5 | 1183 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
