Assay Details
Gene Symbol: Nnat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053601.2 | NP_446053.1 | 2 - 3 | 277 | 63 | |
NR_073089.1 | 3 - 4 | 365 | 63 | |||
GenBank mRNA | CB711943.1 | - | 2 - 3 | 213 | 63 | |
FQ213121.1 | - | 2 - 3 | 235 | 63 | ||
U08290.1 | - | 2 - 3 | 196 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
