Assay Details
Gene Symbol: Cbfa2t2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168542.1 | NP_001162014.1 | 2 - 3 | 293 | 75 | |
XM_006235311.2 | XP_006235373.1 | 2 - 3 | 277 | 75 | ||
XM_006235312.2 | XP_006235374.1 | 3 - 4 | 305 | 75 | ||
XM_008762313.1 | XP_008760535.1 | 2 - 3 | 381 | 75 | ||
GenBank mRNA | FQ222452.1 | - | 2 - 3 | 309 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
