Assay Details
Gene Symbol: Calr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022399.2 | NP_071794.1 | 1 - 2 | 154 | 65 | |
GenBank mRNA | BC062395.1 | - | 1 - 2 | 131 | 5623178 | 65 |
BU671467.1 | - | 1 - 2 | 154 | 65 | ||
D78308.1 | - | 1 - 2 | 106 | 65 | ||
FQ210587.1 | - | 159 | 65 | |||
FQ220096.1 | - | 1 - 2 | 160 | 65 | ||
X53363.1 | - | 1 - 2 | 126 | 65 | ||
X79327.1 | - | 1 - 2 | 104 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
