Assay Details
Gene Symbol: Arpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106919.1 | NP_001100389.1 | 6 - 7 | 580 | 88 | |
XM_006245165.1 | XP_006245227.1 | 5 - 6 | 718 | 88 | ||
XM_006245166.1 | XP_006245228.1 | 6 - 7 | 535 | 88 | ||
GenBank mRNA | FQ214133.1 | - | 6 - 7 | 540 | 88 | |
FQ231947.1 | - | 3 - 4 | 333 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
