Assay Details
Gene Symbol: Arpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106919.1 | NP_001100389.1 | 9 - 10 | 888 | 89 | |
XM_006245165.1 | XP_006245227.1 | 8 - 9 | 1026 | 89 | ||
XM_006245166.1 | XP_006245228.1 | 9 - 10 | 843 | 89 | ||
GenBank mRNA | FQ214133.1 | - | 9 - 10 | 849 | 89 | |
FQ226423.1 | - | 3 - 4 | 288 | 89 | ||
FQ231947.1 | - | 6 - 7 | 641 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
