Assay Details
Gene Symbol: Lox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017061.2 | NP_058757.1 | 7 - 8 | 1437 | 95 | |
XM_006254713.2 | XP_006254775.1 | 5 - 6 | 1520 | 95 | ||
XM_006254714.2 | XP_006254776.1 | 6 - 7 | 1420 | 95 | ||
XM_006254715.2 | XP_006254777.1 | 5 - 6 | 1521 | 95 | ||
GenBank mRNA | BC078861.1 | - | 7 - 8 | 1437 | 7113042 | 95 |
FQ231737.1 | - | 4 - 5 | 461 | 95 | ||
U11038.1 | - | 5 - 6 | 1405 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P038414 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231567 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363428 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738930 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651940 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710535 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911235 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819175 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276622 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62546 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501448 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26626 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88948 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560665 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661741 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166100 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531611 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855095 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867175 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187677 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134347 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393713 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158657 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76551 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718278 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859698 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899059 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529563 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668191 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74458 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230445 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285659 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717290 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938998 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468054 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523825 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517907 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261217 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821226 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311465 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461580 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58042 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33217 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167589 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114599 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172502 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P038414 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P038414 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P038414 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P434724 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P585695 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 702240115 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 84029 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 38526 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 8518 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 54975 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 74849 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 702240115 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10804474 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10804473 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10804469 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10804472 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10804467 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10804475 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10804471 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10804470 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10804464 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10804465 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10804466 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10804468 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10804463 | - | - |
Rat Genome 230 2.0 Array | 1368172_a_at | - | - |
Rat Genome 230 2.0 Array | 1368171_at | - | - |
Rat Genome 230 2.0 Array | 1368172_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1368171_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368172_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1368171_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368171_at | - | - |
Affymetrix HT Rat Focus Array | 1368172_a_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1800010130.rn.1 | TC1800001428.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1800010132.rn.1 | TC1800001428.rn.1 | RTA TC |
Rat Expression Set 230 | 1368171_PM_at | - | - |
Rat Expression Set 230 | 1368171_at | - | - |
Rat Expression Set 230 | 1368172_a_at | - | - |
Rat Expression Set 230 | 1368172_PM_a_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10804463 | - | - |
Rat Genome U34 Set | rc_AI234060_s_at | - | - |
Rat Genome U34 Set | S77494_s_at | - | - |
Rat Genome U34 Set | rc_AA875582_at | - | - |
Rat Genome U34 Set | S66184_s_at | - | - |
Rat Genome U34 Set | R47040_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22167730 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376823 | - | - |
