Assay Details
Gene Symbol: Pgm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017033.2 | NP_058729.2 | 6 - 7 | 1091 | 110 | |
XM_006238442.2 | XP_006238504.1 | 7 - 8 | 1282 | 110 | ||
XM_008763853.1 | XP_008762075.1 | 6 - 7 | 1634 | 110 | ||
GenBank mRNA | BC099807.1 | - | 6 - 7 | 1091 | 7937943 | 110 |
BC128703.1 | - | 6 - 7 | 1096 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
