Assay Details
Gene Symbol: Napa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080585.1 | NP_542152.1 | 2 - 3 | 227 | 60 | |
GenBank mRNA | BC063156.1 | - | 2 - 3 | 286 | 6921187 | 60 |
FQ213217.1 | - | 2 - 3 | 231 | 60 | ||
X89968.1 | - | 2 - 3 | 227 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
