Assay Details
Gene Symbol: Naa20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108595.1 | NP_001102065.1 | 1 - 2 | 116 | 98 | |
XM_006235155.2 | XP_006235217.1 | 1 - 2 | 115 | 98 | ||
GenBank mRNA | BC167020.1 | - | 1 - 2 | 76 | 9025632 | 98 |
FQ234868.1 | - | 1 - 2 | 71 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
