Assay Details
Gene Symbol: Gltscr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106888.1 | NP_001100358.1 | 8 - 9 | 2175 | 65 | |
XM_006244508.2 | XP_006244570.1 | 7 - 8 | 3913 | 65 | ||
XM_008766846.1 | XP_008765068.1 | 9 - 10 | 2237 | 65 | ||
XM_008766847.1 | XP_008765069.1 | 8 - 9 | 2201 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
