Assay Details
Gene Symbol: Pqlc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034952.1 | NP_001030124.1 | 4 - 5 | 523 | 65 | |
GenBank mRNA | BC081879.1 | - | 3 - 4 | 500 | 65 | |
CK477351.1 | - | 4 - 5 | 346 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
